Anti-UNC45A Rabbit Polyclonal Antibody

Supplier: Proteintech
19564-1-AP-150UL
10081-324EA 402 USD
10081-324
Anti-UNC45A Rabbit Polyclonal Antibody
Antibodies
UNC45A, also named as SMAP1, acts as co-chaperone for HSP90. UNC45A prevents the stimulation of HSP90AB1 ATPase activity by AHSA1. It is a positive factor in promoting PGR function in the cell. It is necessary for proper folding of myosin and necessary for normal cell proliferation, normal myotube formation and myosin accumulation during muscle cell development. UNC45A plays a role in erythropoiesis in stroma cells in the spleen. The antibody recognizes the C-term of UNC45A.

Western Blot: Mouse ovary Tissue, 1:500-1:5000; IHC: Human Stomach Tissue, 1:20-1:200

Type: Primary
Antigen: UNC45A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR