Anti-UBASH3B Rabbit Polyclonal Antibody

Supplier: Proteintech
19563-1-AP-150UL
10095-416EA 402 USD
10095-416
Anti-UBASH3B Rabbit Polyclonal Antibody
Antibodies
UBASH3B, also named STS1 or TULA-2, is a protein that contains a ubiquitin-associated domain at the N-terminus, a SH3 domain, and a C-terminal domain with similarities to the catalytic motif of phosphoglycerate mutase. TULA-2 is supposed to form homodimer and interact with ubiquitin and mono-ubiquitinated proteins. TULA-2 functions to promote accumulation of activated target receptors, such as T-cell receptors and EGFR on the cell surface.

Western Blot: Mouse Kidney Tissue, 1:500-1:5000; IHC: Human hepatocirrhosis Tissue, 1:20-1:200; IP: Mouse Kidney Tissue, 1:200-1:1000

Type: Primary
Antigen: UBASH3B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR