Anti-SPHK1 Rabbit Polyclonal Antibody

Supplier: Proteintech
19561-1-AP-100UL
10095-090EA 304.98 USD
10095-090
Anti-SPHK1 Rabbit Polyclonal Antibody
Antibodies
SPHK1, also named as SPHK and SPK, catalyzes the phosphorylation of sphingosine to form sphingosine 1-phosphate (SPP), a lipid mediator with both intra- and extracellular functions. SPHK1 acts on D-erythro-sphingosine and to a lesser extent sphinganine, but not other lipids, such as D,L-threo-dihydrosphingosine, N,N-dimethylsphingosine, diacylglycerol, ceramide, or phosphatidylinositol. The antibody is specific to Ser(311)-phospho of SPHK1(isoform2) or Ser(225)-phospho of SPHK1(isoform1).

Western Blot:L02 Cells, 1:200-1:2000

Type: Primary
Antigen: SPHK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR