Anti-SMTNL1 Rabbit Polyclonal Antibody

Supplier: Proteintech
19560-1-AP-100UL
10094-898EA 304.98 USD
10094-898
Anti-SMTNL1 Rabbit Polyclonal Antibody
Antibodies
Smtnl1 belongs to the smoothelin family. The antibody recognize Ser(301) phospho of Smtn1.

Western Blot: Mouse bladder Tissue, 1:200-1:2000

Type: Primary
Antigen: SMTNL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR