Anti-RUNX1 Rabbit Polyclonal Antibody

Supplier: Proteintech
19555-1-AP-150UL
10094-056EA 402 USD
10094-056
Anti-RUNX1 Rabbit Polyclonal Antibody
Antibodies
RUNX1, also named as AML1 and CBFA2, acts synergistically with ELF4 to transactivate the IL-3 promoter and with ELF2 to transactivate the mouse BLK promoter. RUNX1 inhibits MYST4-dependent transcriptional activation. CBF binds to the core site, 5'-PYGPYGGT-3', of a number of enhancers and promoters, including murine leukemia virus, polyomavirus enhancer, T-cell receptor enhancers, LCK, IL-3 and GM-CSF promoters. The alpha subunit binds DNA and appears to have a role in the development of normal hematopoiesis. Isoform AML-1L interferes with the transactivation activity of RUNX1.

Western Blot: Mouse Liver Tissue, 1:200-1:2000

Type: Primary
Antigen: RUNX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR