Anti-PLK1 Rabbit Polyclonal Antibody

Supplier: Proteintech
19553-1-AP-100UL
10092-470EA 304.98 USD
10092-470
Anti-PLK1 Rabbit Polyclonal Antibody
Antibodies
PLK1, also named as PLK and STPK13, belongs to the protein kinase superfamily, Ser/Thr protein kinase family and CDC5/Polo subfamily. PLK1 is a Serine/threonine-protein kinase that performs several important functions throughout M phase of the cell cycle, including the regulation of centrosome maturation and spindle assembly, the removal of cohesins from chromosome arms, the inactivation of APC/C inhibitors, and the regulation of mitotic exit and cytokinesis. The antibody is specific to Ser(326)-phospho.

Western Blot: MCF-7 Cells, 1:200-1:1000; IHC: Human Colon Tissue, 1:20-1:200

Type: Primary
Antigen: PLK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR