Anti-KIF19 Rabbit Polyclonal Antibody

Supplier: Proteintech
19551-1-AP-150UL
10089-224EA 402 USD
10089-224
Anti-KIF19 Rabbit Polyclonal Antibody
Antibodies
KIF19 belongs to the kinesin-like protein family. The antibody recognizes isoform1 of KIF19 only.

Western Blot: Human Brain Tissue, 1:200-1:2000; IHC: Human Brain, 1:20-1:200

Type: Primary
Antigen: KIF19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR