Anti-MYOF Rabbit Polyclonal Antibody

Supplier: Proteintech
19548-1-AP-150UL
10090-762EA 402 USD
10090-762
Anti-MYOF Rabbit Polyclonal Antibody
Antibodies
MYOF, also named as FER1L3 and KIAA1207, belongs to the ferlin family. MYOF is calcium/phospholipid-binding protein that plays a role in the plasmalemma repair mechanism of endothelial cells that permits rapid resealing of membranes disrupted by mechanical stress. It is involved in endocytic recycling. MYOF implicated in VEGF signal transduction by regulating the levels of the receptor KDR. The antibody can recognize all the 8 isoforms of MYOF.

Western Blot:A549 Cells, 1:500-1:5000; IHC: Human Placenta Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100; FC: HeLa Cells, N/A

Type: Primary
Antigen: MYOF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR