Anti-MXD1 Rabbit Polyclonal Antibody

Supplier: Proteintech
19547-1-AP-150UL
10081-640EA 402 USD
10081-640
Anti-MXD1 Rabbit Polyclonal Antibody
Antibodies
MXD1, also named as MAD, is a transcriptional repressor. MAD1 binds with MAX to form a sequence-specific DNA-binding protein complex which recognizes the core sequence 5'-CAC[GA]TG-3'. MAD thus antagonizes MYC transcriptional activity by competing for MAX.

Western Blot: HepG2 Cells, 1:500-1:5000

Type: Primary
Antigen: MXD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR