Anti-LPIN2 Rabbit Polyclonal Antibody

Supplier: Proteintech
19545-1-AP-150UL
10089-774EA 402 USD
10089-774
Anti-LPIN2 Rabbit Polyclonal Antibody
Antibodies
Lipin 2, also named as KIAA0249, belongs to the lipin family. Defects in LPIN2 are the cause of Majeed syndrome ajeed syndrome which is an autosomal recessive disorder combining features of chronic recurrent multifocal osteomyelitis.

Western Blot: HT-1080 Cells, 1:500-1:5000

Type: Primary
Antigen: LPIN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR