Anti-FEM1B Rabbit Polyclonal Antibody

Supplier: Proteintech
19544-1-AP-150UL
10086-940EA 402 USD
10086-940
Anti-FEM1B Rabbit Polyclonal Antibody
Antibodies
FEM1B, also named as F1AA and KIAA0396, belongs to the fem-1 family. FEM1B is a component of an E3 ubiquitin-protein ligase complex, in which it may act as a substrate recognition subunit. FEM1B is involved in apoptosis by acting as a death receptor-associated protein that mediates apoptosis. And it is involved in glucose homeostasis in pancreatic islet. The antibody recognize the C-term of FEM1B.

Western Blot:PC-3 Cells, 1:200-1:2000

Type: Primary
Antigen: FEM1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR