Anti-FANCD2 Rabbit Polyclonal Antibody

Supplier: Proteintech
19543-1-AP-100UL
10086-856EA 304.98 USD
10086-856
Anti-FANCD2 Rabbit Polyclonal Antibody
Antibodies
FANCD2, also named as FACD and FACD2, is required for maintenance of chromosomal stability. FANCD2 promotes accurate and efficient pairing of homologs during meiosis. FANCD2 is involved in the repair of DNA double-strand breaks, both by homologous recombination and single-strand annealing. It may participate in S phase and G2 phase checkpoint activation upon DNA damage. It promotes BRCA2/FANCD1 loading onto damaged chromatin. FANCD2 may also be involved in B-cell immunoglobulin isotype switching. Defects in FANCD2 are a cause of Fanconi anemia (FA) which is a genetically heterogeneous, autosomal recessive disorder characterized by progressive pancytopenia, a diverse assortment of congenital malformations, and a predisposition to the development of malignancies. The antibody is to phospho-Ser(330) of FANCD2.

Western Blot: HL-60 Cells, 1:200-1:2000; IHC: Human Testis Tissue, 1:20-1:200

Type: Primary
Antigen: FANCD2-phospho(S330)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR