Anti-FANCA Rabbit Polyclonal Antibody

Supplier: Proteintech
19542-1-AP-150UL
10086-850EA 402 USD
10086-850
Anti-FANCA Rabbit Polyclonal Antibody
Antibodies
FANCA, also named as FAA, FACA and FANCH, is DNA repair protein that may operate in a postreplication repair or a cell cycle checkpoint function. FANCA may be involved in interstrand DNA cross-link repair and in the maintenance of normal chromosome stability. Defects in FANCA are a cause of Fanconi anemia (FA) which is a genetically heterogeneous, autosomal recessive disorder characterized by progressive pancytopenia, a diverse assortment of congenital malformations, and a predisposition to the development of malignancies. The antibody is specific to isoform1 of FANCA.

Western Blot: HEK-293 Cells, 1:200-1:2000

Type: Primary
Antigen: FANCA-Specific
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR