Anti-CHKA Rabbit Polyclonal Antibody

Supplier: Proteintech
19537-1-AP-150UL
10082-016EA 402 USD
10082-016
Anti-CHKA Rabbit Polyclonal Antibody
Antibodies
Choline kinase alpha, also named as CHK, CKI and CK, belongs to the choline/ethanolamine kinase family. CHKa may have a regulatory role in phosphatidylcholine synthesis. CHKA catalyze the reaction: ATP + choline = ADP + O-phosphocholine. The human enzyme resembled the rat liver enzyme over the entire sequence. It also resembled the yeast enzyme in the carboxy-terminal region, but not in the amino-terminal region.And it expresses in human brain tissue but not in human skeletal muscle.It has 2 isoforms produced by alternative splicing.The antibody is specific to CHKA.

Western Blot: HepG2 Cells, 1:200-1:1000; IF: HepG2 Cells, 1:10-1:100; IHC: Human breast cancer Tissue, 1:20-1:200

Type: Primary
Antigen: CHKA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR