Anti-SPANXA2/SPANXA1 Rabbit Polyclonal Antibody

Supplier: Proteintech
19527-1-AP-150UL
10095-056EA 402 USD
10095-056
Anti-SPANXA2/SPANXA1 Rabbit Polyclonal Antibody
Antibodies
SPANXA1, also named as SPANXA and SPANXA2, is a member of the SPANX family of cancer/testis-associated genes. It encodes differentially expressed testis-specific proteins that localize to various subcellular compartments. On Western blots of human testis proteins, SPANX antibodies reacts with the bands migrating between 15–20 kDa. The SPANX protein may be a useful target in cancer immunotherapy.

Western Blot:DU 145 Cells, 1:200-1:2000; IF: HEK-293 Cells, 1:10-1:100; IHC: Human skin cancer Tissue, 1:20-1:200

Type: Primary
Antigen: SPANXA2/SPANXA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR