Anti-TICAM2 Rabbit Polyclonal Antibody

Supplier: Proteintech
19524-1-AP-150UL
10095-976EA 402 USD
10095-976
Anti-TICAM2 Rabbit Polyclonal Antibody
Antibodies
TICAM2, also named as TIRAP3, TIRP and TRAM, is involved in IL1-triggered NF-kappa-B activation, functioning upstream of IRAK1, IRAK2, TRAF6, and IKBKB. Physically bridges TLR4 and TICAM1 and functionally transmits LPS-TRL4 signal to TICAM1. TICAM2 is involved in LPS-TLR4 signaling to regulate the MYD88-independent pathway during the innate immune response to LPS.In WB, the MW of TICAM2 is about 27-32kd.

Western Blot:PC-3 Cells, 1:200-1:2000; IHC: Human Testis Tissue, 1:20-1:200

Type: Primary
Antigen: TICAM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR