Anti-ATP6V1G3 Rabbit Polyclonal Antibody

Supplier: Proteintech
19523-1-AP-150UL
10083-382EA 402 USD
10083-382
Anti-ATP6V1G3 Rabbit Polyclonal Antibody
Antibodies
ATP6V1G3,also named as ATP6G3, belongs to the V-ATPase G subunit family. ATP6V1G3 is a catalytic subunit of the peripheral V1 complex of vacuolar ATPase (V-ATPase). V-ATPase is responsible for acidifying a variety of intracellular compartments in eukaryotic cells.

Western Blot: HEK-293 Cells, 1:200-1:2000; IHC: Human Kidney, 1:20-1:200

Type: Primary
Antigen: ATP6V1G3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR