Anti-CES7 Rabbit Polyclonal Antibody

Supplier: Proteintech
19520-1-AP-150UL
10084-680EA 402 USD
10084-680
Anti-CES7 Rabbit Polyclonal Antibody
Antibodies
CES7, also named as CAUXIN, CES4C1, CES5 and FLJ31547, belongs to the type-B carboxylesterase/lipase family. CES7 is involved in the detoxification of xenobiotics and in the activation of ester and amide prodrugs. CES7 catalyze the reaction: A carboxylic ester + H2O = an alcohol + a carboxylate. The antibody can recognize the isoform1 and isoform2.

Western Blot: HepG2 Cells, 1:200-1:1000

Type: Primary
Antigen: CES7
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR