Anti-TXNRD3 Rabbit Polyclonal Antibody

Supplier: Proteintech
19517-1-AP-150UL
10096-544EA 402 USD
10096-544
Anti-TXNRD3 Rabbit Polyclonal Antibody
Antibodies
TXNRD3, also named as TGR and TRXR3, belongs to the class-I pyridine nucleotide-disulfide oxidoreductase family. TXNRD3 displays thioredoxin reductase, glutaredoxin and glutathione reductase activities. TXNRD3 catalyzes the reaction: Thioredoxin + NADP+ = thioredoxin disulfide + NADPH. TXNRD3 promotes disulfide bond formation between GPX4 and various sperm proteins and may play a role in sperm maturation by promoting formation of sperm structural components.

Western Blot: Human Testis Tissue, 1:200-1:2000; IHC: Human Testis, 1:20-1:200

Type: Primary
Antigen: TXNRD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR