Anti-MOGAT2 Rabbit Polyclonal Antibody

Supplier: Proteintech
19514-1-AP-150UL
10081-634EA 402 USD
10081-634
Anti-MOGAT2 Rabbit Polyclonal Antibody
Antibodies
MOGAT2, also named as DC5 and DGAT2L5, belongs to the diacylglycerol acyltransferase family. MOGAT2 catalyze the reaction: Acyl-CoA + 2-acylglycerol = CoA + diacylglycerol. MOGAT2 has a preference toward monoacylglycerols containing unsaturated fatty acids. MOGAT2 plays a central role in absorption of dietary fat in the small intestine by catalyzing the resynthesis of triacylglycerol in enterocytes. And it plays a role in diet-induced obesity. The antibody can recognize all these 3 isoform of MOGAT2.

Western Blot: HEK-293 Cells, 1:200-1:2000

Type: Primary
Antigen: MOGAT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR