Anti-MRAP Rabbit Polyclonal Antibody

Supplier: Proteintech
19512-1-AP-150UL
10090-404EA 402 USD
10090-404
Anti-MRAP Rabbit Polyclonal Antibody
Antibodies
MRAP, also named as C21orf61, FALP and B27, is required for MC2R expression in certain cell types. MRAP is involved in the processing, trafficking or function of MC2R. MRAP may be involved in the intracellular trafficking pathways in adipocyte cells. Mutation of MRAP will cause glucocorticoid deficiency type 2 (GCCD2) which known as familial glucocorticoid deficiency type 2 (FGD2). The antibody can recognize the isoform1,isoform2 and isoform4.

Western Blot: Human Kidney Tissue, 1:200-1:2000; IHC: Human Testis, 1:20-1:200

Type: Primary
Antigen: MRAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR