Anti-TOR2A Rabbit Polyclonal Antibody

Supplier: Proteintech
19511-1-AP-150UL
10096-206EA 402 USD
10096-206
Anti-TOR2A Rabbit Polyclonal Antibody
Antibodies
TOR2A, also named as TORP1, UNQ6408/PRO21181, HEMBA1005096 and PSEC0218, belongs to the clpA/clpB family and Torsin subfamily. TOR2A contain two peptides Salusin-alpha and Salusin-beta. Salusins -alpha and -beta may be endocrine and/or paracrine factors able to increase intracellular calcium concentrations and induce cell mitogenesis. Salusins may also be potent hypotensive peptides.

Western Blot: Mouse Brain Tissue, 1:200-1:2000; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: TOR2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR