Anti-LPA3 Rabbit Polyclonal Antibody

Supplier: Proteintech
19509-1-AP-150UL
10086-262EA 402 USD
10086-262
Anti-LPA3 Rabbit Polyclonal Antibody
Antibodies
LPAR3, also named as EDG7 and LPA3, belongs to the G-protein coupled receptor 1 family. LPAR3 is a receptor for lysophosphatidic acid (LPA), a mediator of diverse cellular activities. LPAR3 may play a role in the development of ovarian cancer. Seems to be coupled to the G(i)/G(o) and G(q) families of heteromeric G proteins. The antibody is specific to LPAR3.

Western Blot:PC-3 Cells, 1:200-1:2000; IHC: Human Heart Tissue, 1:20-1:200

Type: Primary
Antigen: LPA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR