Anti-PACS2 Rabbit Polyclonal Antibody

Supplier: Proteintech
19508-1-AP-150UL
10091-722EA 402 USD
10091-722
Anti-PACS2 Rabbit Polyclonal Antibody
Antibodies
PACS2, also named PACS1L or KIAA0602, is one of PACS (phosphofurin acidic cluster sorting) proteins which are one such family of multifunctional membrane traffic regulators that mediate organ homoeostasis and have important roles in diverse pathologies and disease states. PACS2 is a multifunctional sorting protein that controls the endoplasmic reticulum (ER)-mitochondria communication, ER homeostasis, and apoptosis. PACS2 may be involved in ion channel trafficking, directing acidic cluster-containing ion channels to distinct subcellular compartments.

Western Blot: Mouse Brain Tissue, 1:200-1:2000; IF: MCF-7 Cells, 1:10-1:100

Type: Primary
Antigen: PACS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR