Anti-MAST3 Rabbit Polyclonal Antibody

Supplier: Proteintech
19507-1-AP-150UL
10089-650EA 524.48 USD
10089-650
Anti-MAST3 Rabbit Polyclonal Antibody
Antibodies
MAST3, also named as KIAA0561, belongs to the protein kinase superfamily and AGC Ser/Thr protein kinase family. MAST3 catalyze the reaction: ATP + a protein = ADP + a phosphoprotein.

Western Blot:Jurkat Cells, 1:500-1:5000; IHC: Human Kidney Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: MAST3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR