Anti-ATP9A Rabbit Polyclonal Antibody

Supplier: Proteintech
19504-1-AP-150UL
10083-386EA 402 USD
10083-386
Anti-ATP9A Rabbit Polyclonal Antibody
Antibodies
ATP9A, also named as ATPIIA and KIAA0611, belongs to the cation transport ATPase (P-type) family and Type IV subfamily. ATP9A catalyzes the reaction: ATP + H2O + phospholipid(In) = ADP + phosphate + phospholipid(Out).

Western Blot: Mouse spleen Tissue, 1:500-1:5000

Type: Primary
Antigen: ATP9A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR