Anti-SULT1A3 Rabbit Polyclonal Antibody

Supplier: Proteintech
19499-1-AP-150UL
10095-380EA 402 USD
10095-380
Anti-SULT1A3 Rabbit Polyclonal Antibody
Antibodies
SULT1A3, also named as STM and SULT1A4, belongs to the sulfotransferase 1 family. SULT1A3 catalyzes the sulfate conjugation of phenolic monoamines (neurotransmitters such as dopamine, norepinephrine and serotonin) and phenolic and catechol drugs.

Western Blot:A549 Cells, 1:200-1:1000; IHC: Human hepatocirrhosis Tissue, 1:20-1:200

Type: Primary
Antigen: SULT1A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR