Anti-LRMP Rabbit Polyclonal Antibody

Supplier: Proteintech
19498-1-AP-150UL
10089-788EA 402 USD
10089-788
Anti-LRMP Rabbit Polyclonal Antibody
Antibodies
LRMP, also named as JAW1, is a lymphoid restricted integral membrane protein that is localised to the cytoplasmic face of the endoplasmic reticulum.

Western Blot: Raji Cells, 1:500-1:5000; IF: HepG2 Cells, 1:10-1:100; IHC: Human spleen Tissue, 1:20-1:200

Type: Primary
Antigen: LRMP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR