Anti-AFM Rabbit Polyclonal Antibody

Supplier: Proteintech
19494-1-AP-150UL
10082-584EA 402 USD
10082-584
Anti-AFM Rabbit Polyclonal Antibody
Antibodies
AFM (Afamin) is a member of the albumin superfamily, which comprises albumin, vitamin D-binding protein, a-fetoprotein, and afamin. AFM is present in the plasma/serum, cerebrospinal fluid, and follicular fluid, and functions as a vitamin E-binding protein. Alterations of expression level of AFM have been linked to variety of diseases like cancer.

Western Blot: Human Blood Tissue, 1:500-1:5000

Type: Primary
Antigen: AFM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR