Anti-VLDLR Rabbit Polyclonal Antibody

Supplier: Proteintech
19493-1-AP-150UL
10096-984EA 402 USD
10096-984
Anti-VLDLR Rabbit Polyclonal Antibody
Antibodies
VLDLR binds VLDL and transports it into cells by endocytosis. In order to be internalized, the receptor-ligand complexes must first cluster into clathrin-coated pits. VLDLR binding to Reelin induces tyrosine phosphorylation of Dab1 and modulation of Tau phosphorylation.(cytoplasmic domain of low density ipoprotein receptor). For glycosylated, the MW of VLDLR migrates to ~160kd.

Western Blot: MCF7 Cells, 1:500-1:5000

Type: Primary
Antigen: VLDLR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR