Anti-KIAA0831 Rabbit Polyclonal Antibody

Supplier: Proteintech
19491-1-AP-150UL
10089-198EA 402 USD
10089-198
Anti-KIAA0831 Rabbit Polyclonal Antibody
Antibodies
Barkor, also named as KIAA0831, Atg14 and Atg14L, is required for both basal and inducible autophagy. It plays a role in autophagosome formation and MAP1LC3/LC3 conjugation to phosphatidylethanolamine. Barkor promotes BECN1 translocation from the trans-Golgi network to autophagosomes. This antibody is specific to KIAA0831(Barkor).

Western Blot: Mouse Brain Tissue, 1:200-1:2000

Type: Primary
Antigen: KIAA0831
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR