Anti-ANO8 Rabbit Polyclonal Antibody

Supplier: Proteintech
19485-1-AP-150UL
10082-944EA 402 USD
10082-944
Anti-ANO8 Rabbit Polyclonal Antibody
Antibodies
ANO8, also named as KIAA1623 and TMEM16H, belongs to the anoctamin family. It may act as a calcium-activated chloride channel.

Western Blot: Mouse Brain Tissue, 1:200-1:2000; IF:SH-SY5Y Cells, 1:10-1:100

Type: Primary
Antigen: ANO8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR