Anti-Nestin Rabbit Polyclonal Antibody

Supplier: Proteintech
19483-1-AP-150UL
10091-038EA 402 USD
10091-038
Anti-Nestin Rabbit Polyclonal Antibody
Antibodies
Nestin, also named as NES and Nbla00170, is a class VI intermediate filament protein expressed in stem cells of the central nervous system (CNS) but not in mature CNS cells. Its expression is used extensively as a marker for neural lineage cells derived from human embryonic stem (hES) and induced pluripotent stem (iPS) cells. Nestin is also expressed in non-neural stem cell populations such as pancreatic islet progenitors and hematopoietic progenitors.

Western Blot:rat Brain Tissue, 1:200-1:2000; IHC: Human gliomas Tissue, 1:20-1:200; IF:SH-SY5Y Cells, 1:20-1:200

Type: Primary
Antigen: Nestin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR