Anti-TRPC1 Rabbit Polyclonal Antibody

Supplier: Proteintech
19482-1-AP-150UL
10095-912EA 402 USD
10095-912
Anti-TRPC1 Rabbit Polyclonal Antibody
Antibodies
TRPC1, also named as TRP1, belongs to the transient receptor family and STrpC subfamily. TRPC1 is thought to form a receptor-activated non-selective calcium permeant cation channel. It has been reported that the mGluR1 evoked slow EPSC is mediated by the TRPC1 cation channel. It is operated by a phosphatidylinositol second messenger system activated by receptor tyrosine kinases or G-protein coupled receptors.

Western Blot:PC-3 Cells, 1:500-1:5000

Type: Primary
Antigen: TRPC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR