Anti-PIH1D1 Rabbit Polyclonal Antibody

Supplier: Proteintech
19427-1-AP-150UL
10092-264EA 402 USD
10092-264
Anti-PIH1D1 Rabbit Polyclonal Antibody
Antibodies
PIH1D1 (PIH1 domain-containing protein 1; also known as PIH1 or NOP17), is a ortholog of the yeast nucleolar protein NOP17. PIH1D1 is a conserved subunit of the R2TP complex participating in the assembly of snoRNP and RNA polymerase II complexes. PIH1D1 may also function as a novel modulator of apoptosis pathway.

Western Blot: HeLa Cells, 1:1000-1:10000; IHC: Human lung, 1:20-1:200; IF: Hela, 1:10-1:100

Type: Primary
Antigen: PIH1D1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR