Anti-CHCHD2 Rabbit Polyclonal Antibody

Supplier: Proteintech
19424-1-AP-150UL
10084-426EA 402 USD
10084-426
Anti-CHCHD2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HT-1080 Cells, 1:200-1:2000; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: CHCHD2
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR