Anti-C19orf43 Rabbit Polyclonal Antibody

Supplier: Proteintech
19420-1-AP-150UL
10083-790EA 402 USD
10083-790
Anti-C19orf43 Rabbit Polyclonal Antibody
Antibodies
C19orf43, also named as MGC2803 and fSAP18, is an Uncharacterized protein. It is 176aa,18.4kd. We used the antibody raised from the C19orf43 protein, we get a ~28kd in human placenta, 293 and A431 cell.

Western Blot:A431 Cells, 1:200-1:2000; IHC: Human Heart, 1:20-1:200

Type: Primary
Antigen: C19orf43
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR