Anti-USP10 Rabbit Polyclonal Antibody

Supplier: Proteintech
19374-1-AP-150UL
10096-834EA 402 USD
10096-834
Anti-USP10 Rabbit Polyclonal Antibody
Antibodies
USP10, also named as USP32 and NY-REN-60, belongs to the peptidase C19 family.The deubiquitinating protease USP10 has been identified as a new regulator of p53 in the DNA damage response and tumor development.

Western Blot: HeLa Cells, 1:500-1:5000; IF: Hela, 1:10-1:100

Type: Primary
Antigen: USP10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR