Anti-NUP85 Rabbit Polyclonal Antibody

Supplier: Proteintech
19370-1-AP-150UL
10091-476EA 402 USD
10091-476
Anti-NUP85 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse lung Tissue, 1:500-1:5000; IHC: Human Stomach Tissue, 1:20-1:200; IF: HTERT-RPE1 Cells, 1:20-1:200

Type: Primary
Antigen: NUP85
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR