Anti-EGFL7 Rabbit Polyclonal Antibody

Supplier: Proteintech
19291-1-AP-150UL
10086-306EA 402 USD
10086-306
Anti-EGFL7 Rabbit Polyclonal Antibody
Antibodies
Epidermal growth factor-like domain-containing protein 7 (EGFL7), also known as vascular endothelial statin, is an endothelial cell-derived secreted factor that regulates vascular tube formation. This protein may be involved in the growth and proliferation of tumor cells. Recent studies have reported elevated expression of EGFL7 in several tumors and cancer cell lines, including kidney tumors, malignant gliomas, hepatocellular carcinomas, and colon cancers.

Western Blot: Human Heart Tissue, 1:200-1:2000; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: EGFL7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR