Anti-OAS2 Rabbit Polyclonal Antibody

Supplier: Proteintech
19279-1-AP-150UL
10091-516EA 402 USD
10091-516
Anti-OAS2 Rabbit Polyclonal Antibody
Antibodies
The 2-prime,5-prime oligoadenylate synthetases (OASs), such as OAS2, are interferon-induced proteins characterized by their capacity to catalyze the synthesis of 2-prime,5-prime oligomers of adenosine (2-5As). OAS2 is also named as p69 OAS / p71 OAS. It has 3 isoforms(71/69/20 kDa) produced by alternative splicing. The OAS2 isozymes are 69–71 kDa and form dimers. Glycosylation is essential for its activity. Constitutive expression of the 69-kDa form of the OAS2 protein in human cells inhibited the replication of encephalomyocarditis virus (EMCV) but not that of VSV, Sendai virus, or reovirus. This antibody is specific to isoform p71 and isoform p69 of OAS2.

Western Blot:Jurkat Cells, 1:500-1:5000; IHC: Human breast cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: OAS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR