Anti-CLCA2 Rabbit Polyclonal Antibody

Supplier: Proteintech
19273-1-AP-150UL
10084-836EA 524.48 USD
10084-836
Anti-CLCA2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse lung Tissue, 1:200-1:2000; IHC: Human oesophagus Tissue, 1:20-1:200

Type: Primary
Antigen: CLCA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR