Anti-CDC42EP4 Rabbit Polyclonal Antibody

Supplier: Proteintech
19262-1-AP-150UL
10084-488EA 402 USD
10084-488
Anti-CDC42EP4 Rabbit Polyclonal Antibody
Antibodies
Western Blot:K-562 Cells, 1:500-1:5000

Type: Primary
Antigen: CDC42EP4
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR