Anti-SEMA3C Rabbit Polyclonal Antibody

Supplier: Proteintech
19242-1-AP-150UL
10094-288EA 402 USD
10094-288
Anti-SEMA3C Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Heart Tissue, 1:500-1:5000

Type: Primary
Antigen: SEMA3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR