Anti-TMEM27 Rabbit Polyclonal Antibody

Supplier: Proteintech
19207-1-AP-150UL
10081-170EA 402 USD
10081-170
Anti-TMEM27 Rabbit Polyclonal Antibody
Antibodies
TMEM27(Transmembrane protein 27) is also named as collectrin and Belongs to the TMEM27 family .It is a stimulator of beta cell replicationa and a membrane protein cleaved and shed by pancreatic beta cells that has been proposed as a beta cell mass biomarker.Western blot analysis of mouse collecting duct cell lines detected Tmem27 at an apparent molecular mass of 32 kD(11278314).

Western Blot: Mouse pancreas Tissue, 1:500-1:5000; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: TMEM27
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR