Anti-WDR45 Rabbit Polyclonal Antibody

Supplier: Proteintech
19194-1-AP-150UL
10097-110EA 524.48 USD
10097-110
Anti-WDR45 Rabbit Polyclonal Antibody
Antibodies
IHC: Human Liver, 1:20-1:200; IF: Hela Cells, 1:10-1:100

Type: Primary
Antigen: WDR45
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR