Anti-PSMB5 Rabbit Polyclonal Antibody

Supplier: Proteintech
19178-1-AP-150UL
10091-882EA 524.48 USD
10091-882
Anti-PSMB5 Rabbit Polyclonal Antibody
Antibodies
PSMB5(Proteasome subunit beta type-5) is also named as LMPX, MB1, X and belongs to the peptidase T1B family.The proteasome is a multicatalytic proteinase complex which is characterized by its ability to cleave peptides with Arg, Phe, Tyr, Leu, and Glu adjacent to the leaving group at neutral or slightly basic pH.RNAi-mediated silencing of PSMB5 expression resulted in restoration of bortezomib sensitivity in resistant cells.It has 2 isoforms produced by alternative splicing.

Western Blot:L02 Cells, 1:500-1:5000; IHC: Human breast cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:20-1:200

Type: Primary
Antigen: PSMB5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR