Anti-VPS11 Rabbit Polyclonal Antibody

Supplier: Proteintech
19140-1-AP-150UL
10097-002EA 402 USD
10097-002
Anti-VPS11 Rabbit Polyclonal Antibody
Antibodies
Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. In yeast, Vps proteins are involved in the trafficking of endocytic and biosynthetic proteins to the vacuole, which functionally resembles the lysosome of higher organisms. VPS11 is the human homolog of the yeast class C Vps11 protein, a subunit of HOPS (homotypic fusion and protein transport) complex. Mammalian Vps11 may play a role in vesicle-mediated protein trafficking to lysosomal compartments and in membrane docking/fusion reactions of late endosomes/lysosomes.

Western Blot: Human Brain Tissue, 1:500-1:5000; IF: HepG2 Cells, 1:10-1:100; FC: HepG2 Cells, N/A

Type: Primary
Antigen: VPS11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR