Anti-FOXO6 Rabbit Polyclonal Antibody

Supplier: Proteintech
19122-1-AP-150UL
10087-126EA 402 USD
10087-126
Anti-FOXO6 Rabbit Polyclonal Antibody
Antibodies
FOXO6 is a member of the murine forkhead family of transcription factors. These forkhead transcription factors may play a role in maintenance. FOXO6 mRNA is predominantly expressed in the developing brain, indicating that FOXO6 may play an important role in the nervous system.

Western Blot: Mouse Brain Tissue, 1:200-1:2000; IHC: Human gliomas, 1:20-1:200

Type: Primary
Antigen: FOXO6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR