Anti-GOLPH3 Rabbit Polyclonal Antibody

Supplier: Proteintech
19112-1-AP-150UL
10087-270EA 537.7 USD
10087-270
Anti-GOLPH3 Rabbit Polyclonal Antibody
Antibodies
GOLPH3 (also called GPP34, GMx33, MIDAS, or yeast Vps74p) is a 34-kDa Golgi-associated protein conserved from yeast to human. GOLPH3 binds to PtdIns(4)P-rich trans-Golgi membranes and MYO18A conveying a tensile force required for efficient tubule and vesicle formation . GOLPH3 has been recently demonstrated as a novel oncoprotein amplified in various types of human malignancies, including melanoma, breast, non-small cell lung cancer, gliomas and connective tissue tumors . Enhanced activation of mTOR signalling represents a molecular basis for the oncogenic activity of GOLPH3 .

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human Placenta Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: GOLPH3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR